siRNA Details
Gene Symbol: MYCNUT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125783.1 | 1 | 246 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000433810.1 | - | 1 | 246 | Chr. 2 - 15920399 - 15936017 [+] on Build GRCh38 |
