siRNA Details
Gene Symbol: LOC101928327 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130782.1 | 2 | 334 | |||
GenBank mRNA | BC021184.1 | - | 2 | 339 | ||
BC090895.1 | - | 2 | 307 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000456163.1 | - | 2 | 334 | Chr. 2 - 216870772 - 216871639 [+] on Build GRCh38 |
