siRNA Details
| Gene Symbol: OSER1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038337.2 | 3 | 1337 | |||
| NR_038338.2 | 3 | 1206 | ||||
| NR_038339.2 | 3 | 1049 | ||||
| GenBank mRNA | BC019858.1 | - | 2 | 1090 | ||
| BC032458.1 | - | 3 | 997 | |||
| BC048277.1 | - | 3 | 1049 | |||
| BC067866.1 | - | 3 | 1006 | |||
| BC094781.1 | - | 3 | 1006 | |||

OSER1-AS1
Genomic Map