siRNA Details
Gene Symbol: LINC01014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046786.1 | 3 | 489 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000455307.5 | - | 3 | 489 | Chr. 3 - 178419201 - 178457305 [+] on Build GRCh38 |
