siRNA Details
| Gene Symbol: NPAS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC035412.1 | - | 1 | 508 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000452364.1 | - | 1 | 508 | Chr. 2 - 100993676 - 101002244 [-] on Build GRCh38 | |

NPAS2
Genomic Map