siRNA Details
| Gene Symbol: ROR1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110666.1 | 6 | 1503 | |||
| GenBank mRNA | BC040909.1 | - | 6 | 1503 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000412349.6 | - | 6 | 1503 | Chr. 1 - 64094442 - 64112205 [-] on Build GRCh38 | |

ROR1-AS1
Genomic Map