siRNA Details
| Gene Symbol: ROR1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110665.1 | 3 | 265 | |||
| NR_110666.1 | 4 | 620 | ||||
| GenBank mRNA | AK096291.1 | - | 4 | 354 | ||
| BC040909.1 | - | 4 | 620 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ROR1-AS1| Gene Symbol: ROR1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110665.1 | 3 | 265 | |||
| NR_110666.1 | 4 | 620 | ||||
| GenBank mRNA | AK096291.1 | - | 4 | 354 | ||
| BC040909.1 | - | 4 | 620 | |||