siRNA Details
| Gene Symbol: SMCR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131243.1 | 1 | 50 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000456090.2 | - | 1 | 50 | Chr. 17 - 17674026 - 17677688 [-] on Build GRCh38 | |

SMCR2
Genomic Map