siRNA Details
| Gene Symbol: LOC101927420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136252.1 | 2 | 766 | |||
| NR_136253.1 | 2 | 390 | ||||
| GenBank mRNA | AA527254.1 | - | 2 | 316 | ||
| BC035377.1 | - | 2 | 731 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927420| Gene Symbol: LOC101927420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136252.1 | 2 | 766 | |||
| NR_136253.1 | 2 | 390 | ||||
| GenBank mRNA | AA527254.1 | - | 2 | 316 | ||
| BC035377.1 | - | 2 | 731 | |||