siRNA Details
| Gene Symbol: LOC101929282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110246.1 | 1, 2 | 227 | |||
| NR_110247.1 | 1, 2 | 227 | ||||
| GenBank mRNA | BX100556.1 | - | 1, 2 | 57 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929282| Gene Symbol: LOC101929282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110246.1 | 1, 2 | 227 | |||
| NR_110247.1 | 1, 2 | 227 | ||||
| GenBank mRNA | BX100556.1 | - | 1, 2 | 57 | ||