siRNA Details
| Gene Symbol: LOC102724714 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135567.1 | 2 | 326 | |||
| GenBank mRNA | AW969319.1 | - | 2 | 326 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000429730.1 | - | 2 | 381 | Chr. 2 - 207662375 - 207667024 [+] on Build GRCh38 | |

LOC102724714
Genomic Map