siRNA Details
| Gene Symbol: HCG14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104117.1 | 2 | 251 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000444986.1 | - | 2 | 251 | Chr. 6 - 28896530 - 28897322 [+] on Build GRCh38 | |

HCG14
Genomic Map