siRNA Details
| Gene Symbol: LINC00570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047499.1 | 3 | 1037 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000417473.6 | - | 3 | 1037 | Chr. 2 - 11393981 - 11403077 [+] on Build GRCh38 | |

LINC00570
Genomic Map