siRNA Details
| Gene Symbol: MIR3681HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110196.1 | 6 | 1518 | |||
| GenBank mRNA | AK001558.1 | - | 6 | 1506 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000412294.5 | - | 6 | 1506 | Chr. 2 - 12007128 - 12578348 [+] on Build GRCh38 | |

MIR3681HG
Genomic Map