siRNA Details
| Gene Symbol: ATMIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK302556.1 | - | 3 | 1679 | ||
| Gene Symbol: LOC105374301 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017007603.1 | XP_016863092.1 | 2 | 2613 | ||
| XM_017007604.1 | XP_016863093.1 | 2 | 2557 | |||
| XM_017007605.1 | XP_016863094.1 | 2 | 2578 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000423193.1 | - | 2 | 477 | Chr. 3 - 857124 - 858091 [+] on Build GRCh38 | |

ATMIN
Genomic Map