siRNA Details
| Gene Symbol: LINC00403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120392.1 | 4 | 661 | |||
| NR_120394.1 | 2 | 502 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000421423.1 | - | 2 | 502 | Chr. 13 - 112107119 - 112108015 [+] on Build GRCh38 | |

LINC00403
Genomic Map