siRNA Details
Gene Symbol: LOC284930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122046.1 | 9 | 1534 | |||
GenBank mRNA | AK093107.1 | - | 9 | 1505 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000423737.5 | - | 9 | 1534 | Chr. 22 - 47631674 - 47855600 [+] on Build GRCh38 |
