siRNA Details
| Gene Symbol: LOC284930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122046.1 | 5 | 696 | |||
| GenBank mRNA | AK093107.1 | - | 5 | 667 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000423737.5 | - | 5 | 696 | Chr. 22 - 47631674 - 47855600 [+] on Build GRCh38 | |

LOC284930
Genomic Map