siRNA Details
| Gene Symbol: SLC25A5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028443.1 | 1 | 2380 | |||
| NR_134914.1 | 2 | 1299 | ||||
| NR_134915.1 | 2 | 1167 | ||||
| GenBank mRNA | AK092094.1 | - | 1 | 2356 | ||
| AK094280.1 | - | 2 | 1167 | |||
| BC028211.1 | - | 2 | 1299 | |||
| BC050320.1 | - | 1 | 2337 | |||

SLC25A5-AS1
Genomic Map