siRNA Details
| Gene Symbol: SLC25A5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028443.1 | 1 | 2343 | |||
| NR_134914.1 | 2 | 1262 | ||||
| NR_134915.1 | 2 | 1130 | ||||
| GenBank mRNA | AK092094.1 | - | 1 | 2319 | ||
| AK094280.1 | - | 2 | 1130 | |||
| BC028211.1 | - | 2 | 1262 | |||
| BC050320.1 | - | 1 | 2300 | |||

SLC25A5-AS1
Genomic Map