siRNA Details
| Gene Symbol: LOC101927501 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110387.1 | 3 | 501 | |||
| GenBank mRNA | AK056105.1 | - | 3 | 501 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000440955.1 | - | 3 | 501 | Chr. X - 43176994 - 43226598 [+] on Build GRCh38 | |

LOC101927501
Genomic Map