siRNA Details
| Gene Symbol: LOC101929331 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121587.1 | 2 | 370 | |||
| GenBank mRNA | DA257806.1 | - | 2 | 370 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929331| Gene Symbol: LOC101929331 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121587.1 | 2 | 370 | |||
| GenBank mRNA | DA257806.1 | - | 2 | 370 | ||