siRNA Details
| Gene Symbol: LINC01567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122072.1 | 3 | 925 | |||
| GenBank mRNA | BC132761.1 | - | 1 | 134 | ||
| BC132789.1 | - | 1 | 134 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000414816.1 | - | 3 | 925 | Chr. 16 - 24661422 - 24671062 [-] on Build GRCh38 | |

LINC01567
Genomic Map