siRNA Details
| Gene Symbol: LOC100147773 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125374.1 | 2 | 1014 | |||
| GenBank mRNA | AK124964.1 | - | 2 | 1014 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000423121.1 | - | 2 | 1014 | Chr. 1 - 165768929 - 165775176 [+] on Build GRCh38 | |

LOC100147773
Genomic Map