siRNA Details
| Gene Symbol: LINC00703 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108054.1 | 3 | 660 | |||
| NR_108055.1 | 2 | 557 | ||||
| NR_108056.1 | 2 | 430 | ||||
| GenBank mRNA | AK095699.1 | - | 2 | 557 | ||
| DA500285.1 | - | 3 | 485 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00703| Gene Symbol: LINC00703 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108054.1 | 3 | 660 | |||
| NR_108055.1 | 2 | 557 | ||||
| NR_108056.1 | 2 | 430 | ||||
| GenBank mRNA | AK095699.1 | - | 2 | 557 | ||
| DA500285.1 | - | 3 | 485 | |||