siRNA Details
| Gene Symbol: LINC01280 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110392.1 | 4 | 1044 | |||
| GenBank mRNA | AK307257.1 | - | 4 | 1044 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000449783.1 | - | 4 | 1044 | Chr. 2 - 216590426 - 216606938 [-] on Build GRCh38 | |

LINC01280
Genomic Map