siRNA Details
| Gene Symbol: STK24-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF339799.1 | - | 2 | 735 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000434547.1 | - | 2 | 735 | Chr. 13 - 98577244 - 98578830 [+] on Build GRCh38 | |

STK24-AS1
Genomic Map