siRNA Details
| Gene Symbol: LOC101929662 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120670.1 | 3 | 2151 | |||
| GenBank mRNA | BC041821.1 | - | 3 | 2146 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000435367.2 | - | 3 | 2151 | Chr. 10 - 85432863 - 85448961 [+] on Build GRCh38 | |

LOC101929662
Genomic Map