siRNA Details
| Gene Symbol: LINC01087 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108087.1 | 2 | 2908 | |||
| GenBank mRNA | BC036114.1 | - | 2 | 2905 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000431979.1 | - | 2 | 2908 | Chr. 2 - 131637025 - 131649615 [+] on Build GRCh38 | |

LINC01087
Genomic Map