siRNA Details
Gene Symbol: LINC01087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108087.1 | 2 | 2347 | |||
GenBank mRNA | BC036114.1 | - | 2 | 2344 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000431979.1 | - | 2 | 2347 | Chr. 2 - 131637025 - 131649615 [+] on Build GRCh38 |
