siRNA Details
| Gene Symbol: SH3BP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA843106.1 | - | 1 | 171 | ||
| BX114836.1 | - | 1 | 268 | |||
| CA429664.1 | - | 1 | 662 | |||
| Gene Symbol: SH3BP5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046084.1 | 1 | 820 | |||
| GenBank mRNA | AL133111.1 | - | 1 | 820 | ||

SH3BP5
Genomic Map