siRNA Details
Gene Symbol: LOC101929312 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110610.1 | 2 | 329 | |||
GenBank mRNA | BC043288.2 | - | 2 | 295 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101929312 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110610.1 | 2 | 329 | |||
GenBank mRNA | BC043288.2 | - | 2 | 295 |