siRNA Details
| Gene Symbol: LOC101928887 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134456.1 | 2 | 2151 | |||
| GenBank mRNA | BC050028.1 | - | 2 | 2155 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000425902.2 | - | 2 | 2151 | Chr. 10 - 64901136 - 64924518 [-] on Build GRCh38 | |

LOC101928887
Genomic Map