siRNA Details
| Gene Symbol: LOC284577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK095633.1 | - | 3 | 583 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000442318.1 | - | 3 | 712 | Chr. 1 - 205455929 - 205469024 [+] on Build GRCh38 | |

LOC284577
Genomic Map