siRNA Details
Gene Symbol: LOC101928751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125964.1 | 2 | 1143 | |||
GenBank mRNA | BC031108.1 | - | 2 | 1143 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000457217.1 | - | 2 | 1143 | Chr. 1 - 175877343 - 175880468 [+] on Build GRCh38 |
