siRNA Details
| Gene Symbol: LOC101928751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125964.1 | 3 | 1703 | |||
| GenBank mRNA | BC031108.1 | - | 3 | 1703 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000457217.1 | - | 3 | 1703 | Chr. 1 - 175877343 - 175880468 [+] on Build GRCh38 | |

LOC101928751
Genomic Map