siRNA Details
| Gene Symbol: ATP2B2-IT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046766.1 | 2 | 282 | |||
| GenBank mRNA | BX110985.1 | - | 2 | 303 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000419591.1 | - | 2 | 282 | Chr. 3 - 10626021 - 10626807 [-] on Build GRCh38 | |

ATP2B2-IT2
Genomic Map