siRNA Details
Gene Symbol: ATP2B2-IT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046766.1 | 1 | 22 | |||
GenBank mRNA | BX110985.1 | - | 1 | 43 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000419591.1 | - | 1 | 22 | Chr. 3 - 10626021 - 10626807 [-] on Build GRCh38 |
