siRNA Details
| Gene Symbol: LOC101928298 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120636.1 | 1, 2 | 74 | |||
| GenBank mRNA | BC032914.2 | - | 1, 2 | 74 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000429539.1 | - | 1, 2 | 74 | Chr. 10 - 10058722 - 10063502 [+] on Build GRCh38 | |

LOC101928298
Genomic Map