siRNA Details
Gene Symbol: LOC101928298 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120636.1 | 4 | 481 | |||
GenBank mRNA | BC032914.2 | - | 4 | 481 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000429539.1 | - | 4 | 481 | Chr. 10 - 10058722 - 10063502 [+] on Build GRCh38 |
