siRNA Details
| Gene Symbol: LINC00853 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047498.1 | 2 | 625 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000429328.2 | - | 2 | 625 | Chr. 1 - 47179250 - 47180339 [+] on Build GRCh38 | |

LINC00853
Genomic Map