siRNA Details
Gene Symbol: LINC00853 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047498.1 | 1 | 155 | |||
GenBank mRNA | AI792909.1 | - | 1 | 155 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000429328.2 | - | 1 | 155 | Chr. 1 - 47179250 - 47180339 [+] on Build GRCh38 |
