siRNA Details
Gene Symbol: LOC101927869 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109963.1 | 2 | 236 | |||
GenBank mRNA | BC043533.1 | - | 2 | 236 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101927869 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109963.1 | 2 | 236 | |||
GenBank mRNA | BC043533.1 | - | 2 | 236 |