siRNA Details
Gene Symbol: LOC101928782 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110836.1 | 2 | 419 | |||
GenBank mRNA | AK098603.1 | - | 2 | 419 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000415393.1 | - | 2 | 419 | Chr. 7 - 131910220 - 131948953 [+] on Build GRCh38 |
