siRNA Details
| Gene Symbol: POT1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125718.1 | 10 | 1426 | |||
| NR_125719.1 | 7 | 738 | ||||
| GenBank mRNA | AK090675.1 | - | 4 | 371 | ||
| BC142949.1 | - | 10 | 1382 | |||
| BX648695.1 | - | 7 | 697 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

POT1-AS1| Gene Symbol: POT1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125718.1 | 10 | 1426 | |||
| NR_125719.1 | 7 | 738 | ||||
| GenBank mRNA | AK090675.1 | - | 4 | 371 | ||
| BC142949.1 | - | 10 | 1382 | |||
| BX648695.1 | - | 7 | 697 | |||