siRNA Details
| Gene Symbol: POT1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125718.1 | 11 | 2583 | |||
| NR_125719.1 | 8 | 1895 | ||||
| GenBank mRNA | BC142949.1 | - | 11 | 2538 | ||
| BX648695.1 | - | 8 | 1851 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

POT1-AS1| Gene Symbol: POT1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125718.1 | 11 | 2583 | |||
| NR_125719.1 | 8 | 1895 | ||||
| GenBank mRNA | BC142949.1 | - | 11 | 2538 | ||
| BX648695.1 | - | 8 | 1851 | |||