siRNA Details
| Gene Symbol: CCDC103 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BQ448043.1 | - | 1 | 509 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF131813.1 | - | 1 | 1630 | Chr. 17 - 44903051 - 44904819 [-] on Build GRCh38 | |

CCDC103
Genomic Map