siRNA Details
| Gene Symbol: LINC00320 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024090.2 | 8 | 1547 | |||
| NR_109786.1 | 8 | 1658 | ||||
| NR_109787.1 | 7 | 1423 | ||||
| NR_109788.1 | 7 | 1484 | ||||
| GenBank mRNA | AK094858.1 | - | 7 | 1372 | ||
| BC042856.1 | - | 8 | 1502 | |||
| BC044727.1 | - | 8 | 1502 | |||
| BC047405.1 | - | 8 | 1620 | |||
| BX537735.1 | - | 7 | 1484 | |||

LINC00320
Genomic Map