siRNA Details
| Gene Symbol: LINC00184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_033927.1 | 2 | 1323 | |||
| GenBank mRNA | AJ421679.1 | - | LIT1706 HANC |
1 | 827 | |
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000429269.1 | - | 2 | 1323 | Chr. 1 - 234629311 - 234634780 [+] on Build GRCh38 | |

LINC00184
Genomic Map