siRNA Details
Gene Symbol: LINC00184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_033927.1 | 2 | 1262 | |||
GenBank mRNA | AJ421679.1 | - | LIT1706 HANC |
1 | 766 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000429269.1 | - | 2 | 1262 | Chr. 1 - 234629311 - 234634780 [+] on Build GRCh38 |
